Merck
BAH500250309
SP Bel-Art® Glove Box Bellows Type Gloves for 8 in. Glove Ports
latex, size 9 medium
Synonym(e):
glove box
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Materialien
latex
Größe
9 medium
Leistungsmerkmale
to hold Bellows and Gloves
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H50025-0309
Allgemeine Beschreibung
Made from natural latex with a removable hand, the sleeves fit 20cm (8") circular ports and 18 x 25cm (7 x 10") oval ports. Use with H50025-0320 Clamping Rings to hold gloves in place.
Leistungsmerkmale und Vorteile
  • Forms a leak-proof unit with O-ring construction integral with the box
  • Bellows allows cooling air circulation
  • When the glove and sleeve are separated, the sleeve remains attached to the box
  • Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
  • Length of the extended sleeve and glove is approximately 76cm (30")
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)