Wichtige Dokumente
289515
Thiabendazol
98%
Synonym(e):
2-(4-Thiazolyl)-benzimidazol
Alle Fotos(1)
About This Item
Empirische Formel (Hill-System):
C10H7N3S
CAS-Nummer:
Molekulargewicht:
201.25
Beilstein:
611403
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352005
PubChem Substanz-ID:
Empfohlene Produkte
Assay
98%
Form
solid
SMILES String
c1ccc2[nH]c(nc2c1)-c3cscn3
InChI
1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)
InChIKey
WJCNZQLZVWNLKY-UHFFFAOYSA-N
Signalwort
Warning
H-Sätze
P-Sätze
Gefahreneinstufungen
Aquatic Acute 1 - Aquatic Chronic 1
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Miin-Huey Lee et al.
International journal of food microbiology, 150(2-3), 157-163 (2011-08-27)
Penicillium digitatum causes green mold on citrus, resulting in severe postharvest fruit decay and economic losses in many citrus-producing areas of the world. Forty isolates of P. digitatum were cultured from citrus groves, packinghouses, and local markets in Taiwan, and
Andrea Peano et al.
Veterinary dermatology, 23(2), 131-135 (2012-02-09)
In this study, we evaluated the antifungal susceptibility of Malassezia pachydermatis to clotrimazole (CTZ), miconazole (MCZ), and thiabendazole (TBD), azole derivatives employed in aural formulations labeled for treatment of canine otitis. The procedure for in vitro testing was based on
Hye Ji Cha et al.
PLoS biology, 10(8), e1001379-e1001379 (2012-08-29)
Studies in diverse organisms have revealed a surprising depth to the evolutionary conservation of genetic modules. For example, a systematic analysis of such conserved modules has recently shown that genes in yeast that maintain cell walls have been repurposed in
P Fonderie et al.
Parasitology, 139(10), 1301-1308 (2012-06-22)
Studies on anthelmintic resistance in equine parasites do not include facultative parasites. Halicephalobus gingivalis is a free-living bacterivorous nematode and a known facultative parasite of horses with a strong indication of some form of tolerance to common anthelmintic drugs. This
Matthieu Groh et al.
Clinical and experimental rheumatology, 30(1 Suppl 70), S107-S109 (2012-07-26)
Polyarteritis nodosa (PAN), a systemic necrotising vasculitis that affects medium- and small-sized arteries, has visceral involvement in 40-60% of the patients. According to the Five-Factor Score (FFS), it is associated with poor outcome. We describe a patient who underwent orthotopic