Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13541

Supelco

Diaion® WA30 free base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

form

solid

Quality Level

packaging

pkg of 100 g

parameter

100 °C max. temp.

moisture

~49%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (highly porous)

particle size

16-50 mesh

operating pH

0-9

capacity

1.5 meq/mL by wetted bed volume
3.0 meq/g by dry weight

Looking for similar products? Visit Product Comparison Guide

Application

Diaion® WA30 free base is an anion exchange resin for high molecular weight organic acids. It has been used in conjunction with electrodialysis to remove salt and selective organic acids from salted Japanese apricot.
Weakly basic anion exchange resin that is best in class for high MW organic acids. It also has a strong decolorization capability. Applications for the resin include: water treatment, pretreatment of corn syrup, beet sugar and dextrose, refining of formalin, glycerine, and enzymes, and as a weak base catalyst. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Diaion is a registered trademark of Mitsubishi Chemical Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W Takatsuji et al.
Journal of bioscience and bioengineering, 88(3), 348-351 (2005-10-20)
With the aim of repeatedly reusing the solution used to season salted ume (Japanese apricot), we investigated its desalting and deacidification by electrodialysis, precipitation, and adsorption. Although NaCl and acids could be easily removed from used seasoning solution by electrodialysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service