Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

216569

Supelco

Amberlite IRA-410 Ion Exchange Resin

chloride form, 20-25 mesh, matrix styrene/divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

product name

Amberlite IRA-410 chloride form, chloride form, matrix styrene/divinylbenzene, 20-25 mesh

vapor density

<1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

form

beads

autoignition temp.

800 °F

parameter

40 °C OH- form max. temp.
77 °C Cl- form max. temp.

moisture

45-51%

technique(s)

LPLC: suitable

matrix

styrene/divinylbenzene

matrix active group

benzyldimethyl(2-hydroxyethyl)ammonium functional group

particle size

20-25 mesh
600-750 μm

operating pH

0-14

capacity

1.25 meq/mL by wetted bed volume

separation technique

anion exchange

InChI

1S/C13H20NO.C10H10.ClH/c1-4-12-7-5-6-8-13(12)11-14(2,3)9-10-15;1-3-9-7-5-6-8-10(9)4-2;/h4-8,15H,1,9-11H2,2-3H3;3-8H,1-2H2;1H/q+1;;/p-1

InChI key

DPRMXNMXCFHCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite IRA-410, anion exchange resin, is an effective catalyst especially for those reactions involving low catalyst concentrations and relatively low temperature. Amberlite IRA-410 is a gel-type resin having styrene-divinyl-benzene skeletal structure.

Application

Amberlite IRA-410 was used as packing material for columns. Amberlite IRA-410 was also used in urinary analysis by HPLC.
Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.

Features and Benefits

Strongly basic gel-type resin with benzyldimethyl(2-hydroxyethyl)ammonium functionality chloride ion form useful in catalytic applications.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatyana Elkin et al.
Materials (Basel, Switzerland), 12(4) (2019-02-20)
Polystyrene-b-polyethylene glycol (PS-b-PEG) amphiphilic block copolymers featuring a terminal tridentate N,N,N-ligand (terpyridine) were synthesized for the first time through an efficient route. In this approach, telechelic chain-end modified polystyrenes were produced via reversible addition-fragmentation chain-transfer (RAFT) polymerization by using terpyridine
Determination of selenium by atomic absorption spectrometry with simultaneous retention of selenium (IV) and tetrahydroborate (III) on an anion-exchange resin followed by flow injection hydride generation from the solid phase
Carrero, PabloaE, and JulianaF Tyson.
Analyst, 122, 915-919 (1997)
Catalysis by Anion Exchange Resins.
Schmidle, Claude J., and Richard C. Mansfield.
Ind. and Eng. Chem., 44, 1388-1390 (1952)
I R Sequeira et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 24(9), e401-e411 (2012-07-05)
We explored the temporal dynamics of the lactulose mannitol test and the influence of a single dose of aspirin. Twenty healthy female volunteers each received 600 mg aspirin or placebo in random sequence and were subsequently dosed with 10 g

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service