Skip to Content
MilliporeSigma

Skip To

06428

Amberlite IRC120 Ion Exchange Resin

greener alternative

hydrogen form, 16-50 mesh, strongly acidic

Synonym(s):

Amberlite IR120 hydrogen form

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amberlite IRC120 H, hydrogen form, strongly acidic, hydrogen form

vapor density

>1 (vs air)

Quality Segment

vapor pressure

17 mmHg ( 20 °C)

description

cross-linkage 8%, moisture 53-58%

form

beads

autoignition temp.

800 °F

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

121 °C max. temp.

technique(s)

LPLC: suitable

loss

~50% loss on drying, 110 °C

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid

particle size

620-830 μm

operating pH

0-14

capacity

1.8 meq/mL by wetted bed volume

greener alternative category

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

General description

Amberlite IR120 is a gel-type strongly acidic cation exchange resin and a heterogeneous catalyst.

Application

Strongly acidic cation exchange resin suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc..
Amberlite IR120 has been used as a catalyst support to form chelates with Co2+ or Ru3+ ions for subsequent hydrogen generation by hydrolysis of ammonia borane. It may be used as a catalyst for the oxirane ring-opening reaction effected by hydrogen peroxide (H2O2) during the synthesis of epoxides from triglycerides source. Amberlite IR120 hydrogen form may be used to investigate the ion-exchange equilibria of H+/Pb2+, H+/Ni2+, and H+/Cr3+ systems.

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1032206423216399
separation technique

cation exchange

separation technique

cation exchange

separation technique

cation exchange

separation technique

cation exchange

particle size

620-830 μm

particle size

620-830 μm

particle size

<300 μm

particle size

<300 μm

parameter

121 °C max. temp.

parameter

121 °C max. temp.

parameter

120 °C max. temp.

parameter

120 °C max. temp.

matrix active group

sulfonic acid

matrix active group

sulfonic acid

matrix active group

sulfonic acid functional group

matrix active group

sulfonic acid functional group

matrix

styrene-divinylbenzene (gel)

matrix

styrene-divinylbenzene (gel)

matrix

styrene-divinylbenzene (macroreticular)

matrix

styrene-divinylbenzene (macroreticular)

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. I wonder about the stability of regenerated Amberlite and if there is any recommendation of storage solution for regenerated material or could it be stored in the regeneration solution for example.

    1 answer
    1. This material is a strongly acidic cation exchange resin and stores quite well in the regenerated form. Regeneration should take place with an acid sufficient to inhibit any possible microbial contamination. Following regeneration, rinse with several bed volumes of deionized water prior to storage. Alternatively, a final rinse/storage solution of 20% aqueous ethanol may also be used. Rinse well prior to re-use.

      Do not allow the column to dry out. When rehydrated, dry resins are susceptible to bead breakage due to rapid re-swelling. Should drying occur, rehydrate with a saturated NaCl solution. The high osmotic pressure will minimize rapid re-swelling. The salt can then be removed by successive dilutions to prevent a rapid change in osmotic pressure resulting in bead breakage.

      Helpful?

  2. Salve, vorrei sapere se questa resina è adatta ad essere usata in un deionizzatore. Grazie

    1 answer
    1. This product is not tested for use as a deionizer. This item is assigned a quality level of 100 and is intended for research use only. For more information regarding the quality levels within the M-Clarity program see the link below:
      https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/m-clarity-program

      Helpful?

Reviews

No rating value

Active Filters