Skip to Content
MilliporeSigma

A9960

Amberlite IRA-67 free base

gel form, 16-50 mesh (wet)

Synonym(s):

1,2-bis(ethenyl)benzene,4-[(E)-2-[(E)-2-but-3-enoxyethenoxy]ethenoxy]but-1-ene,N-[3-(dimethylamino)propyl]prop-2-enamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
47101511
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

gel form

Quality Segment

contains

~60% moisture content

parameter

≤60 °C temperature

moisture

~60%

matrix

Acrylic (gel)

matrix active group

polyamine

particle size

16-50 mesh (wet)

pH

0-7(effective)

capacity

1.6 meq/mL, 5.6 meq/g

SMILES string

N(CCCNC(=O)C=C)(C)C.O(CCC=C)\C=C\O\C=C\OCCC=C.c1(c(cccc1)C=C)C=C

InChI

1S/C12H18O3.C10H10.C8H16N2O/c1-3-5-7-13-9-11-15-12-10-14-8-6-4-2;1-3-9-7-5-6-8-10(9)4-2;1-4-8(11)9-6-5-7-10(2)3/h3-4,9-12H,1-2,5-8H2;3-8H,1-2H2;4H,1,5-7H2,2-3H3,(H,9,11)/b11-9+,12-10+;;

InChI key

ZGXNLYRRYYOMNG-JDDKLYJPSA-N

Application

Amberlite IRA-67 free base can be used in anion exchange media. It has been used as an adsorbent for lactic acid extraction from aqueous solution, and for the extraction of rhinacanthin from Rhinacanthus nasutus leaves.
Weakly basic anion exchange resin with an unusually high capacity for large organics. Common applications include: deacidification, deionization of process liquors, isolation of acidic natural products, purification of novabiacin and cephalosporins, separation of neutral and acidic amino acids below pH 10, removal of heparinic acid at low pH, concentration of citric acid, and demineralization of cheese whey.

Other Notes

Previously known as IRA-68.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library