Documents
Z768677
Nalgene® media bottles, style 2019
capacity 2000 mL
Synonym(s):
bottle nalgene, nalgene containers, sterile bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
square bottle
transparent PETG
Joint
Joints threaded neck (53B)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations
with cap
packaging
pack of 12 ea (2 xpack of 6)
manufacturer/tradename
Nalgene® 2019-2000
capacity
2000 mL
General description
  • Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
  • Designed for storing buffers, cultures, and culture media for various laboratory applications.
  • Features a leakproof closure for sample protection.
  • Heat-shrink band around closure and neck facilitate tamper-evident seal.
  • Ideal for refrigerated/frozen storage down to -40°C.
Features and Benefits
  • Avoids depyrogenation, expensive washing, and autoclaving steps
  • Sterile and non-pyrogenic
  • Obtained in low particulate format
  • Validated by extensive documentation
  • Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
  • Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries