Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1552

Sigma-Aldrich

Phytatray

L × W × H 11.4 cm × 8.6 cm × 6.4 cm, PETG, sterile; γ-irradiated

Synonym(s):

Phytatray Plant Culture, Plant Culture Tray, Plant Growth Tray

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161500
NACRES:
NB.22

material

PETG

sterility

sterile; γ-irradiated

L × W × H

11.4 cm × 8.6 cm × 6.4 cm

application(s)

agriculture

General description

These trays are sterile, disposable plant cell culture vessels made of PETG. They are ideal for numerous plant cell culture applications, such as high throughput screening.

Packaging

Packaged with sterile lids and trays/case in each box.

Legal Information

Phytatray is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rodrigo A Gutiérrez et al.
Genome biology, 8(1), R7-R7 (2007-01-16)
Carbon (C) and nitrogen (N) metabolites can regulate gene expression in Arabidopsis thaliana. Here, we use multi-network analysis of microarray data to identify molecular networks regulated by C and N in the Arabidopsis root system. We used the Arabidopsis whole
Viktoriya Coneva et al.
Plant physiology, 175(1), 376-391 (2017-08-11)
Thicker leaves allow plants to grow in water-limited conditions. However, our understanding of the genetic underpinnings of this highly functional leaf shape trait is poor. We used a custom-built confocal profilometer to directly measure leaf thickness in a set of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service