MilliporeSigma
  • Genetic Architecture and Molecular Networks Underlying Leaf Thickness in Desert-Adapted Tomato
Genetic Architecture and Molecular Networks Underlying Leaf Thickness in Desert-Adapted Tomato
Plant physiology (2017-08-11)
Viktoriya Coneva, Margaret H Frank, Maria A de Luis Balaguer, Mao Li, Rosangela Sozzani, Daniel H Chitwood
ABSTRACT
Thicker leaves allow plants to grow in water-limited conditions. However, our understanding of the genetic underpinnings of this highly functional leaf shape trait is poor. We used a custom-built confocal profilometer to directly measure leaf thickness in a set of introgression lines (ILs) derived from the desert tomato
Sigma-Aldrich
Phytatray, L × W × H 11.4 cm × 8.6 cm × 6.4 cm, PETG, sterile; γ-irradiated