Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V8505

Magenta vessel

GA-7

Synonym(s):

Laboratory vessel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

material

polycarbonate body
polypropylene closure

description

Autoclavable

W × L × H

77 mm × 77 mm × 97 mm

application(s)

agriculture

Looking for similar products? Visit Product Comparison Guide

General description

Unit is unbreakable and designed for prolonged use. Unique closure and drip ring make this product ideal for plant tissue culture use.

Application

Magenta vessel is suitable for plant tissue culture, multiplication, transport and pre-transplanting. It has been used for transplanting. It has also been used for microcosm construction and assembling.

Features and Benefits

  • Stimulates fast seed germination and easier inoculation and transfer
  • Provides large relative growing area and ensures low incubation shelving cost
  • Designed for improved stacking and occupies less space in autoclaving, shipping & storage
  • Excellent plant protection - no breakage or leakage
  • Autoclavable and reusable
  • Minimizes contamination and the resulting profit loss
  • Easy operation and less handling when used with MAGENTA 7-Way Tray
  • Lightweight

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thuluz Meza-Menchaca et al.
Microorganisms, 8(6) (2020-06-27)
Mycotoxins from the Fusarium genus are widely known to cause economic losses in crops, as well as high mortalities rates among immunocompromised humans. However, to date, no correlation has been established for the ability of Fusarium to cause cross-kingdom infection
Alexandra K Nothstein et al.
PloS one, 14(4), e0214219-e0214219 (2019-04-20)
Selenium plays an important, but vastly neglected role in human nutrition with a narrow gap between dietary deficiency and toxicity. For a potential biofortification of food with Se, as well as for toxicity-risk assessment in sites contaminated by Se, modelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service