Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7186

Greiner FLUOTRAC 600 384 well plates

flat bottom black polystyrene wells, maximum volume 28 μL, non-sterile

Synonym(s):

384 immunoassay plates, 384 multiwell plates, 384 well microplates, 384 well microtiter plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
black polystyrene
flat bottom black polystyrene wells
polystyrene
size:
384 wells
sterility:
non-sterile
binding type:
high binding surface
feature:
lid: no

material

black polystyrene
flat bottom black polystyrene wells
polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 40

manufacturer/tradename

Greiner 784077

maximum volume

28 μL

size

384 wells

well volume

28 μL

working volume

25 μL

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

FLUOTRAC 600: 384 flat bottom, non-treated microtiter plate with an optimal working volume of 4-25 μL; maximum well capacity of 28 μL.

Other Notes

384 flat well HiBase microtiter plate, small volume, high-binding, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0,15 mm; width: 85.48 +/- 0,15 mm; curvature: approx.150 μm.

Suitability

Black plates minimize background fluorescence. FLUOTRAC 600 binds up to 600 ng/cm2. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

FLUOTRAC is a trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service