Skip to Content
MilliporeSigma

P6116

Nunc® MaxiSorp 384 well plates

384 well plate, white, polystyrene, 120ul/well, pinchbar design, 30/cs, size 384 wells, flat bottom white polystyrene wells, maximum volume 120 μL

Synonym(s):

384 multiwell plate, 384 well microplates, 384 well microtiter plate, 384 well plate, ELISA plate, immunoassay plate, Nunc 460372

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom white polystyrene wells, white polystyrene
Size:
384 wells
Sterility:
non-sterile
Binding type:
(MaxiSorp surface)
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom white polystyrene wells, white polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 30 ea (internal packs of 10)

manufacturer/tradename

Nunc 460372

maximum volume

120 μL

plate L × W

128 mm × 86 mm

size

384 wells

well maximum volume

120 μm

working volume

100 μL

color

white

suitability

suitable for (colorimetric assays, sample storage; luminescence and fluorescence assays, ELISA)

binding type

(MaxiSorp surface)

General description

MaxiSorp plates for ELISA

  • Same footprint as a 96 well plate
  • Working volume range of 10-100 μL
  • Compatible with robotic, automated readers and liquid handling systems
  • Assay miniaturization increases productivity and reduces cost
  • C-well for optical clarity and sample recovery
  • Reduced risk of well-to-well contamination
  • Optimizes robotic access to wells
  • White and black plates minimize crosstalk
  • Rounded square wells eliminate wicking (capillary action)
  • Flat bottom for optical reading
  • Optimized for scintillation counting
  • Black 384 well plate reduces background fluorescence and minimizes backlight scatter
  • White 384 well plate maximizes reflection of signal while maintaining low background
  • Clear 384 well plate for optical clarity for readthrough applications

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nunc® MaxiSorp 384 well plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library