Skip to Content
MilliporeSigma

M6936

Greiner high binding 384 well plates

black polystyrene, flat bottom clear polystyrene wells (with micro-clear bottom), sterile

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, Greiner 781097

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41122107
Material:
black polystyrene , flat bottom clear polystyrene wells (with micro-clear bottom)
Size:
384 wells
Sterility:
sterile
Binding type:
high binding surface
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black polystyrene , flat bottom clear polystyrene wells (with micro-clear bottom)

sterility

sterile

feature

lid: no

packaging

case of 40

manufacturer/tradename

Greiner 781097

maximum volume

131 μL

size

384 wells

working volume

110 μL

binding type

high binding surface

General description

384W microplate,with an optimal working volume of 15-110 μL; maximum well capacity of 131 μL. Growth area: 10 mm(2)
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Application

Black plates minimize background fluorescence. μClear film improves Luminescence; Fluorescence; Cell monitoring; Transmission; and Imaging by reducing background, cross-talk and autofluorescence.

Other Notes

384 flat (F) solid well microtiter plate, high-binding, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0.15 mm; width: 85.48 +/- 0.15 mm; curvature: approx.150 μm.

Legal Information





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library