Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M041130

Sigma-Aldrich

6-Fluorescein Phosphoramidite

configured for ABI

Synonym(s):

6-FAM-phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H58N3O10P
CAS Number:
Molecular Weight:
843.94
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥97% (31P-NMR)
≥97.0% (reversed phase HPLC)

color

solid white to off-white

solubility

acetonitrile: 0.1 M, clear, colorless to faintly yellow (liquid)

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR

compatibility

configured for ABI

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCCNC(=O)c1ccc2C(=O)OC3(c4ccc(OC(=O)C(C)(C)C)cc4Oc5cc(OC(=O)C(C)(C)C)ccc35)c2c1)OCCC#N

InChI

1S/C46H58N3O10P/c1-29(2)49(30(3)4)60(55-25-15-22-47)54-24-14-12-11-13-23-48-40(50)31-16-19-34-37(26-31)46(59-41(34)51)35-20-17-32(56-42(52)44(5,6)7)27-38(35)58-39-28-33(18-21-36(39)46)57-43(53)45(8,9)10/h16-21,26-30H,11-15,23-25H2,1-10H3,(H,48,50)

InChI key

MGPYJVWEJNTXLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoramidite technology is ideally suited for the modification ofsynthetic oligonucleotides with covalently attached reporter moieties,linkers, spacers, or haptens. Sigma-Aldrich provides popular reporters andlinkers ready to use for DNA/RNA synthesis machines such as β-cyanoethylphosphoramidites. 6-Fluorescein Phosphoramidite is one such label whose absorbance maxima is 496 nm and emission maxima is 520 nm.Key Features of 6-Fluorescein Phosphoramidite:

  • 6-Fluorescein Phosphoramidite consists of one isomer derived from6-carboxy fluorescein
  • Couples to the 5′-end of the oligonucleotide using standard synthesisprotocols
  • Protecting groups on the fluorescein moiety are removed under standardconditions of cleavage and deprotection with concentrated ammonia
  • A detritylation step is not required since the fluorescein phosphoramiditedoes not contain a DMT-group
  • The labeled oligonucleotide is ready for use in most applications after theevaporation of ammonia. Standard procedures can be used if additionalpurification is required.
  • Lot-to-lot consistent purity and performance
  • Manufactured under a certified ISO 9001 quality system6-Fluorescein Phosphoramidite is configured for ABI Synthesizers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service