Skip to Content
MilliporeSigma

M043180

6-Hexachloro-Fluorescein Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

6-(4,7,2′,4′,5′,7′-Hexachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H52Cl6N3O10P
CAS Number:
Molecular Weight:
1050.61
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
41116105
MDL number:
eCl@ss:
32120505
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (31P-NMR), ≥97.0% (reversed phase HPLC)

color

white to off-white, powder

suitability

conforms to structure for H-NMR

compatibility

configured for PerkinElmer, configured for Polygen

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCCNC(=O)c1cc(Cl)c2C(=O)OC3(c4cc(Cl)c(OC(=O)C(C)(C)C)c(Cl)c4Oc5c(Cl)c(OC(=O)C(C)(C)C)c(Cl)cc35)c2c1Cl)OCCC#N

InChI

1S/C46H52Cl6N3O10P/c1-23(2)55(24(3)4)66(61-19-15-16-53)60-18-14-12-11-13-17-54-40(56)25-20-28(47)31-32(33(25)50)46(65-41(31)57)26-21-29(48)38(63-42(58)44(5,6)7)34(51)36(26)62-37-27(46)22-30(49)39(35(37)52)64-43(59)45(8,9)10/h20-24H,11-15,17-19H2,1-10H3,(H,54,56)

InChI key

RDZTYXJTMDFAOO-UHFFFAOYSA-N

General description

Phosphoramidite technology is ideally suited for the modification ofsynthetic oligonucleotides with covalently attached reporter moieties,linkers, spacers or haptens. Sigma-Aldrich provides popular reporters andlinkers ready to use for DNA/RNA synthesis machines such as β-cyanoethylphosphoramidites. 6-Hexachloro-Fluorescein Phosphoramidite is one such label whose absorbance maxima 535 nm and emission maxima is 556 nm.6-Hexachloro-Fluorescein Phosphoramidite is configured for Expedite™ and Polygen® Synthesizers.

Legal Information

Expedite is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
PolyGen is a registered trademark of PolyGen GmbH.


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library