Skip to Content
MilliporeSigma
All Photos(1)

Documents

A111000

Sigma-Aldrich

DMT-dA(bz) Phosphoramidite

Synonym(s):

DMT-dA(bz) Amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyadenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H52N7O7P
CAS Number:
Molecular Weight:
857.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)

color

white to off-white

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C47H52N7O7P/c1-32(2)54(33(3)4)62(59-27-13-26-48)61-40-28-42(53-31-51-43-44(49-30-50-45(43)53)52-46(55)34-14-9-7-10-15-34)60-41(40)29-58-47(35-16-11-8-12-17-35,36-18-22-38(56-5)23-19-36)37-20-24-39(57-6)25-21-37/h7-12,14-25,30-33,40-42H,13,27-29H2,1-6H3,(H,49,50,52,55)/t40-,41+,42+,62?/m0/s1

InChI key

GGDNKEQZFSTIMJ-AKWFTNRHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dA(bz) Phosphoramidite belongs to the group of DNA Phosphoramidites.

Its key features include:

  • Exocyclic amine functions are protected by a benzoyl group (dA(bz) anddC(bz)) or isobutyryl group (dG(ib))
  • Recommended cleavage and deprotection conditions are 8 hours at 55 °Cor 24 hours at room temperature using concentrated ammonia solution,for standard base-protected oligonucleotides
  • The high coupling efficiency of Proligo′s DNA phosphoramidites leads tohigh-yield and high-quality oligonucleotides

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service