Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M046080

Sigma-Aldrich

Cyanine 5 Phosphoramidite

configured for Polygen, configured for PerkinElmer

Synonym(s):

2-[5-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H72N4O4P
CAS Number:
Molecular Weight:
944.21
UNSPSC Code:
41116105
NACRES:
NA.21

product line

Proligo Reagents

Assay

≥90% (31P-NMR)
≥95.0% (reversed phase HPLC)

color

clear blue0.1M Acetonitrile

compatibility

configured for PerkinElmer
configured for Polygen

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C1=C(N/2CCCOP(N(C(C)C)C(C)C)OCCC#N)C=CC=C1)(C)C2=C\C=C\C=C\C(C3(C)C)=[N+](CCCOC(C4=CC=CC=C4)(C5=CC=C(OC)C=C5)C6=CC=CC=C6)C7=C3C=CC=C7.[Cl-]

InChI

1S/C60H72N4O4P.ClH/c1-46(2)64(47(3)4)69(67-44-23-40-61)68-45-25-42-63-55-33-22-20-31-53(55)59(7,8)57(63)35-18-12-17-34-56-58(5,6)52-30-19-21-32-54(52)62(56)41-24-43-66-60(48-26-13-10-14-27-48,49-28-15-11-16-29-49)50-36-38-51(65-9)39-37-50;/h10-22,26-39,46-47H,23-25,41-45H2,1-9H3;1H/q+1;/p-1

InChI key

GWCNEDQILVVBDJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cyanine 5 Phosphoramidite (C60H17N4O4P) is a dye with absorbance maxima at 646 nm and emission maxima 662 nm. It has a molecular weight of 888.78. Phosphoramidite technology is ideally suited for the modification ofsynthetic oligonucleotides with covalently attached reporter moieties,linkers, spacers or haptens.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service