M043130
6-Hexachloro-Fluorescein Phosphoramidite
configured for ABI
Synonym(s):
6-(4,7,2′,4′,5′,7′-Hexachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
About This Item
Recommended Products
Assay
≥97% (31P-NMR)
≥97.0% (reversed phase HPLC)
Quality Level
color
white to off-white, powder
suitability
conforms to structure for H-NMR
conforms to structure for H-NMR
compatibility
configured for ABI
storage temp.
−20°C
SMILES string
CC(C)N(C(C)C)P(OCCCCCCNC(=O)c1cc(Cl)c2C(=O)OC3(c4cc(Cl)c(OC(=O)C(C)(C)C)c(Cl)c4Oc5c(Cl)c(OC(=O)C(C)(C)C)c(Cl)cc35)c2c1Cl)OCCC#N
InChI
1S/C46H52Cl6N3O10P/c1-23(2)55(24(3)4)66(61-19-15-16-53)60-18-14-12-11-13-17-54-40(56)25-20-28(47)31-32(33(25)50)46(65-41(31)57)26-21-29(48)38(63-42(58)44(5,6)7)34(51)36(26)62-37-27(46)22-30(49)39(35(37)52)64-43(59)45(8,9)10/h20-24H,11-15,17-19H2,1-10H3,(H,54,56)
InChI key
RDZTYXJTMDFAOO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service