Skip to Content
MilliporeSigma
All Photos(2)

Documents

L5391

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Isolectin B4 (BSI-B4), peroxidase conjugate, lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Bandieraea simplicifolia

Quality Level

conjugate

peroxidase conjugate

Assay

≥90% protein basis (Warburg-Christian)

form

lyophilized powder

potency

<200 μg per mL agglutination activity (using human blood group B erythrocytes)

peroxidase activity

40-160 units/mg protein

composition

Protein, ~95% modified Warburg-Christian

technique(s)

blood typing: suitable
fractionation: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

General Western Blot Protocol:
  • Glycoprotein sample size: 500ng
  • Lectin Concentration: 0.1ug/ml

  1. Load samples at 500 ng of glycoprotein per lane
  2. Run 4-20% Bis-Tris SDS page gel
  3. Transfer gel to a PVDF membrane
  4. Block membrane for 1 hr at RT with RIPA buffer (R0278 Sigma)
  5. Incubate HRP lectin at 0.1ug/ml with RIPA buffer for 2 hours at RT
  6. Wash membrane 5 x 5 minutes with 25ml RIPA buffer
  7. Detect using chemiluminescent substrate (CPS1-120)

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Unit Definition

One unit will form 1 mg purpurogallin in 20 sec from pyrogallol at pH 6.0 at 20 °C.

Physical form

Contains sodium citrate buffer salts and calcium chloride

Preparation Note

Prepared from peroxidase type VI using a modification of the method of O′Sullivan, et al. Repurified by affinity chromatography after conjugation.

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle Y Hamline et al.
Developmental biology, 468(1-2), 110-132 (2020-07-22)
BCOR is a critical regulator of human development. Heterozygous mutations of BCOR in females cause the X-linked developmental disorder Oculofaciocardiodental syndrome (OFCD), and hemizygous mutations of BCOR in males cause gestational lethality. BCOR associates with Polycomb group proteins to form
C Kaur et al.
Archives of histology and cytology, 56(5), 475-484 (1993-12-01)
The present study describes the development and differentiation of microglial cells in the spinal cord of postnatal rats ranging from 1 day to 3 weeks of age. Using the monoclonal antibody OX-42, three different morphological forms of immunoreactive cells (SP
Dariusz Orlowski et al.
Neuromodulation : journal of the International Neuromodulation Society, 20(5), 417-423 (2017-02-22)
The use of Deep Brain Stimulation (DBS) in treatment of various brain disorders is constantly growing; however, the number of studies of the reaction of the brain tissue toward implanted leads is still limited. Therefore, the aim of our study
Christophe Dubois et al.
Journal of the American College of Cardiology, 55(20), 2232-2243 (2010-05-15)
We compared biological repair after acute myocardial infarction (AMI) with selected porcine progenitor cell populations. Cell types and mechanisms responsible for myocardial repair after AMI remain uncertain. In a blinded, randomized study, we infused autologous late-outgrowth endothelial progenitor cells (EPC)
Hamed Zaer et al.
Scientific reports, 10(1), 16223-16223 (2020-10-03)
Stereotactic radiosurgery (SRS) has proven an effective tool for the treatment of brain tumors, arteriovenous malformation, and functional conditions. However, radiation-induced therapeutic effect in viable cells in functional SRS is also suggested. Evaluation of the proposed modulatory effect of irradiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service