Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09468

Sigma-Aldrich

Buffer tablets pH 7.2

for preparing buffer solution acc. to WEISE for staining of blood smears

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.21

Quality Level

form

solid

IVD

for in vitro diagnostic use

pH

7.2 (20 °C in H2O)

bulk density

1000 kg/m3

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

General description

Buffer tablets pH 7.2 - for preparing buffer solution according to Weise for staining of blood smears, are used to prepare buffer solutions according to Weise and pH-buffered staining solutions. Phosphate buffer solutions are used in hematology, for the dilution of staining solutions and for the rinsing steps in the staining process. The buffer tablets acc. to Weise will produce a buffered solution (pH 6.4/pH 6.8/pH 7.2) that stabilizes the staining result and gives reproducible staining results. Buffer solution according to Weise pH 6.4 results in bright-orange stained erythrocytes and differentiated stained cells with nuclei, whereby the erythrocytes are stained orange when using pH 6.8. Buffer solution acc. to Weise pH 7.2 produces grayish stained erythrocytes and intense red-violet stained cells with nuclei.
100 tablets are sufficient for 100 l buffer solution, each produced solution can be stable up to 4 weeks. Buffer tablets according to Weise are IVD products and CE registered. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

pH-value (1 tablet in 1 l of water): 7.15 - 7.25
Crushing strength: ≥ 50 N
Disintegration time: ≤ 20 min
Suitability for blood-smear staining: conforms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service