Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0010-02

Sephadex® G-10

Cytiva 17-0010-02, pack of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

feature

autoclavable In wet form (pH 7.0) at 120°C for 30 min

packaging

pack of 500 g

manufacturer/tradename

Cytiva 17-0010-02

parameter

flow rate (The beads behave as rigid spheres obeying Darcy′s Law.)

matrix

cross-linked dextran

particle size

40-120 μm (dry)
55-165 (wet)

working range

2-13

Related Categories

General description

Sephadex® G-10 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules > 700 molecular weight. Smallest Exlusion limit in the Sephadex® range.

Application

Sephadex® is a gel filtration medium prepared by crosslinking Dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-10 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-50 is available in 4 different particle sizes (Course, Medium, Fine & Superfine) and Superfine has the smallest bead size for higher efficiency fractionation with shorter diffusion distances. Cource and Medium are are preferred for large scale group separations where high flow rates and Low operating pressures are required.

Features and Benefits

  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
  • Suitable for buffer exchange and cleanup of biological samples, for example peptides, small Proteins, and oligosaccharides larger than 700 molecular weight
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
  • Lowest exclusion limit (700 molecular weight) in the Sephadex® G-range

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Protocols

Column Packing and Preparation for Size Exclusion Chromatography

Related Content

Sephadex® from Cytiva and Darcy’s law in Size Exclusion Chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service