Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G50150

Supelco

Sephadex® G-50 Size Exclusion Resin

Medium, 100-285 mesh

Synonym(s):

Dextran 50

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Product Name

Sephadex® G-50, Medium

Quality Level

form

slurry

technique(s)

LPLC: suitable

matrix

Agarose

matrix active group

polymer

swelling

-1 g swells to 9-11 mL

bead size

50-150 μm

separation technique

size exclusion (SEC)

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
Fractionation Range (MW)
Globular Proteins: 1,500 - 30,000
Dextrans: 500 - 10,000

Other Notes

G50150-100G′s updated product number is GE17-0043-01
G50150-500G′s updated product number is GE17-0043-02

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supramolecular organization of S12363-liposomes prepared with two different remote loading processes.
Chemin, C., et al.
Biochimica et Biophysica Acta - Biomembranes, 1788(5), 926-935 (2009)
M E Clark et al.
Molecular and cellular biology, 10(10), 5106-5113 (1990-10-01)
In HeLa cells, RNA polymerase III (pol III)-mediated transcription is severely inhibited by poliovirus infection. This inhibition is due primarily to the reduction in transcriptional activity of the pol III transcription factor TFIIIC in poliovirus-infected cells. However, the specific binding
C K Lee et al.
Journal of virology, 54(3), 731-738 (1985-06-01)
An immunoassay was used to examine the interaction between a herpes simplex virus protein, ICP8, and various types of DNA. The advantage of this assay is that the protein is not subjected to harsh purification procedures. We characterized the binding
P S Kay et al.
Nucleic acids research, 15(4), 1559-1577 (1987-02-25)
An RNA molecule consisting of the 5' exon and intervening sequence (IVS) of Tetrahymena precursor rRNA was oxidized with sodium periodate to convert the ribose moiety of the 3' terminal guanosine into a dialdehyde form. The modified RNA undergoes a
Harvey S Penefsky
FEBS letters, 579(10), 2250-2252 (2005-04-07)
Pi binding by the F(1)-ATPase of beef heart mitochondria and of the Escherichia coli plasma membrane (E. coli F(1)) was examined by two methods: the centrifuge column procedure [Penefsky, H.S. (1977) J. Biol. Chem. 252, 2891-2899] and the Paulus pressure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service