Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0435-01

Column PD-10, Empty

Cytiva 17-0435-01

Synonym(s):

PD-10 Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

packaging

pkg of 50 ea

manufacturer/tradename

Cytiva 17-0435-01

column I.D.

15 mm

Related Categories

General description

Empty Disposable PD-10 Columns are economical, plastic columns for single-use applications using gravity flow. The columns hold up to approx. 13 mL of chromatography media. The frits can be used as an upper bed support that prevents the medium from running dry under gravity flow.

Together with LabMate Buffer Reservoir, up to 25 mL of buffer and/or sample can be applied, which reduces handling time considerably.

Features and Benefits

  • Economical and versatile columns for gravity flow applications.
  • Empty PD-10 columns can be packed with a wide range of bulk media.
  • Easy to use.
  • Use PD-10 Columns with LabMate Buffer Reservoir for convenient equilibration, sample application, and wash.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service