Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

G25300

Sigma-Aldrich

Sephadex® G-25 Size Exclusion Resin

Coarse, 50-140 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Product Name

Sephadex® G-25, Coarse

Quality Level

100
200

technique(s)

LPLC: suitable

swelling

1 g swells to 4-6 mL gel

bead size

100-300 μm

InChI

1S/C6H12O6.C3H8O3/c7-1-2-3(8)4(9)5(10)6(11)12-2;4-1-3(6)2-5/h2-11H,1H2;3-6H,1-2H2/t2-,3-,4-,5-,6-;/m1./s1

InChI key

BCPVZFFJGCURNR-OCOFDJSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

G25300-100G′s updated product number is GE17-0034-01
G25300-500G′s updated product number is GE17-0034-02

Application

Sephadex® G-25 is a gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography. Sephadex® G-25 has been used to study the acceleration of diabetic wound healing and the treatment of epithelial damage.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of trypsin treatment on rabbit muscle phosphofructokinase.
K Emerk et al.
Archives of biochemistry and biophysics, 164(1), 233-240 (1974-09-01)
J C Fredenburgh et al.
The Journal of biological chemistry, 267(36), 26150-26156 (1992-12-25)
Plasminogen, the zymogen form of the fibrinolytic enzyme plasmin, is known to undergo plasmin-mediated modification in vitro. The modified form, Lys-plasminogen, is superior to the native Glu-plasminogen in fibrin binding and as a substrate for activation by tissue-type plasminogen activator
E Berman et al.
Carbohydrate research, 113(1), 141-150 (1983-02-16)
As a prelude to studies using bovine N-acetylglucosaminide-beta-(1 leads to 4)-galactosyltransferase to label membrane-surface glycoproteins with isotopically enriched D-galactose, the structural specificity of the enzymic reaction with water-soluble, hen ovalbumin has been examined. The enzyme-catalyzed transfer of D-galactose from UDP-D-galactose
P Kilzer et al.
Microvascular research, 30(3), 270-285 (1985-11-01)
A new method is described for assessing changes in vascular permeation by albumin in multiple tissues of the same animal in response to intravascular injection of vasoactive agents. Following intravenous injection of 51Cr-RBC, 125I-BSA, and 57Co-EDTA, a test substance (i.e.
B P Hills et al.
International journal of food microbiology, 36(2-3), 187-198 (1997-05-20)
The survival of Salmonella Typhimurium LT2 in randomly packed beds of glass beads, microporous silica particles and Sephadex microspheres is examined. It is shown that the decrease in the percentage cell recovery in these porous materials at reduced water content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service