Documents
CLS430591
Corning® non-treated culture dishes
D × H 100 mm × 20 mm
Synonym(s):
cell culture dishes, cell culture plates, culture dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
material
polystyrene
round
sterility
sterile
packaging
sleeve of 20
manufacturer/tradename
Corning 430591
D × H
100 mm × 20 mm
surface area
55 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachment, for applications where cell attachment is not desired
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
Visit the Document Library
Kristina Xiao Liang et al.
Experimental neurology, 337, 113536-113536 (2020-12-03)
The inability to reliably replicate mitochondrial DNA (mtDNA) by mitochondrial DNA polymerase gamma (POLG) leads to a subset of common mitochondrial diseases associated with neuronal death and depletion of neuronal mtDNA. Defining disease mechanisms in neurons remains difficult due to