Skip to Content
MilliporeSigma

P5606

Petri dishes, polystyrene

size 100 mm × 20 mm , nominal size

Synonym(s):

petri dish, petri plate, petri plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.14
UNSPSC Code:
41121812
Material:
circles , polystyrene
Sterility:
sterile; γ-irradiated
Binding type:
non-treated surface
Packaging:
pack of 400 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

circles , polystyrene

sterility

sterile; γ-irradiated

feature

disposable, lid, Optically clear with consistently flat bottoms

packaging

pack of 400 ea

dish capacity

~150 mL

dish diam.

100 mm

size

100 mm × 20 mm , nominal size

binding type

non-treated surface

General description

Designed for easy one hand opening and closing, edges of tops and bottoms are squared off to provide a firm sure grip. Flare and length of lid skirt allows lid to be removed and replaced easily. Dishes have three venting ribs on the underside of the lid to prevent condensation build up.

  • Standard slippable petri dish
  • Clear top and bottom viewing surfaces
  • Disposable (single use) polystyrene petri dish for general microbiological use
  • Molded from medical-grade virgin polystyrene
  • Optically clear
  • Radiation sterilized
  • Contents sterile if sleeve is unopened
  • Ventilation ribs allow for free air circulation and reduce condensation during incubation
  • Flared lid skirt and squared corners for easy one-hand operation, especially with gloves

Please note: Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner; this results in a size difference in the outer diameter of approx 10 mm.


Still not finding the right product?

Explore all of our products under Petri dishes, polystyrene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library