Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
circles , polystyrene
Sterility:
sterile; γ-irradiated
Binding type:
non-treated surface
Packaging:
pack of 500 ea
material
circles , polystyrene
sterility
sterile; γ-irradiated
feature
disposable, lid, beveled stacking rings recommended foruse with automated plate filling and streaking systems
packaging
pack of 500 ea
technique(s)
cell culture: suitable
dish capacity
~115 mL
dish diam.
100 mm
size
, 100 mm × 15 mm , nominal size
binding type
non-treated surface
General description
Designed for easy one hand opening and closing, edges of tops and bottoms are squared off to provide a firm sure grip. Flare and length of lid skirt allows lid to be removed and replaced easily. Dishes have three venting ribs on the underside of the lid to prevent condensation build up.
Please note: Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner; this results in a size difference in the outer diameter of approx 10 mm.
- Standard slippable petri dish
- Clear top and bottom viewing surfaces
- Disposable (single use) polystyrene petri dish for general microbiological use
- Molded from medical-grade virgin polystyrene
- Optically clear
- Radiation sterilized
- Contents sterile if sleeve is unopened
- Ventilation ribs allow for free air circulation and reduce condensation during incubation
- Flared lid skirt and squared corners for easy one-hand operation, especially with gloves
Please note: Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner; this results in a size difference in the outer diameter of approx 10 mm.
Still not finding the right product?
Explore all of our products under Petri dishes, polystyrene
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.