Key Documents
CLS430588
Corning® non-treated culture dishes
D × H 35 mm × 10 mm
Synonym(s):
cell culture dishes, cell culture plates, culture dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
material
polystyrene
round
sterility
sterile
usage
1.6-2.4 mL (Optimal culture medium volume)
packaging
sleeve of 20
manufacturer/tradename
Corning 430588
D × H
35 mm × 10 mm
surface area
8 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachment, for applications where cell attachment is not desired
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.