Key Documents
CLS3959
Corning® 96 well Polypropylene Deep Well Plate
round bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
polypropylene
polypropylene plate
round bottom clear
translucent bottom
well volume:
1 mL
wells:
96 wells , storage block
material
polypropylene
polypropylene plate
round bottom clear
translucent bottom
sterility
non-sterile
feature
lid: no
skirt
manufacturer/tradename
Corning 3959
packaging
bag of 5 ×
well volume
1 mL
wells
96 wells , storage block
binding type
non-treated surface
General description
Corning offers a complete polypropylene storage system consisting of plates, blocks, and mats.
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)