Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPDW11CS

Deep well plate

size 96 wells, size 1.1 mL, pkg of 10x5plates/cs, sterile

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear bottom
round bottom wells
size:
1.1 mL
96 wells

material

clear bottom
round bottom wells

sterility

sterile

feature

lid: no
skirt

manufacturer/tradename

Axygen Inc P-DW-11-C-S
Corning

packaging

pkg of 10x5plates/cs

size

1.1 mL
96 wells

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service