MilliporeSigma
All Photos(4)
Key Documents
CLS3956
Corning® 96 well Polypropylene Deep Well Plate
V-bottom clear, polypropylene, bag of 10 ×, sterile, lid: no
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
well volume:
0.5 mL
wells:
96 wells , storage block
material
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
sterility
sterile
feature
lid: no
skirt
manufacturer/tradename
Corning 3956
packaging
bag of 10 ×
well volume
0.5 mL
wells
96 wells , storage block
capacity
0.5 mL
binding type
non-treated surface
General description
Corning offers a complete polypropylene storage system consisting of plates, blocks, and mats.
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)