Key Documents
BR701362
BRAND® mat cover for deep well plate
volume 2.2 mL
Synonym(s):
Accessories, Cover mats, Deep-well Plates, Sample Storage
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
sterility
non-sterile
manufacturer/tradename
BRAND 701362
volume
2.2 mL
General description
BRAND® mat cover is an ethylene vinyl acetate sealing mat for 96 deep well plates with square-shaped wells. Sealing mats are ideal for short-term storage and reliably protect samples against contamination and evaporation.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)