MilliporeSigma
All Photos(4)
Key Documents
CLS3960
Corning® 96 well Polypropylene Deep Well Plate
V-bottom clear, polypropylene, bag of 5 ×, sterile, lid: no
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
size:
96 wells
material
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
sterility
sterile
feature
lid: no
skirt
manufacturer/tradename
Corning 3960
packaging
bag of 5 ×
size
96 wells
well volume
2 mL
wells
96 wells , storage block
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
Corning offers a complete polypropylene storage system consisting of plates, blocks, and mats.
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.
Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.
Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.
Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.