Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A373-C6

97011321, human skin, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

A373-C6, 97011321

biological source

human skin

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

IL-1 receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human malignant melanoma IL-1 sensitive

Cell Line Description

The cell line A375-C6 has been shown to be sensitive to the IL-1 mediated anti-proliferative effect. It has been derived from the parental line A375 (Sigma Catalogue number. 88113005) by limiting dilution together with its twin clone A375-C5 (Sigma Catalogue number. 97011320) which is IL-1 unresponsive. A375-C6 could be useful for a growth inhibition assay for IL-1. The cells have been eradicated from mycoplasma at ECACC.

Application

IL-1 growth inhibition assay, IL-1 receptor analysis

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 11
D16S539: 9
D5S818: 12
D7S820: 9
THO1: 8
TPOX: 8,10
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 i.e. seeding at 1-3 x 10,000 cells/cm2using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service