Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LA-N-2

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 6041202, human nerve, Large elongated cells + small round cells in clusters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human nerve

packaging

tube of 5 μg 06041202-DNA-5UG
pkg of vial of cells 06041202-1VL

growth mode

Not specified

karyotype

Modal no. 73, range 46 - 73

morphology

Large elongated cells + small round cells in clusters

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Neuroblastoma

Cell Line Description

Established by Seeger et al., (1977) from the primary site of a noncatecholamine-producing neuroblastoma of a 3-year-old female with clinical Stage IV neuroblastoma. The culture consists mostly of large, elongated cells with processes with some small round cells adhering to the elongated cells. The cells are tumourigenic in nude mice.

Culture Medium

EMEM (with non-essential amino acids) and Ham′s F12 (1:1 mixture) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

For routine maintenance, split cultures after they have become dense i.e. once every 3-4 weeks at a 1:20 to 1:50 ratio; 8% CO2; 37oC. The culture consists of large elongated cells with processes and small round cells. The latter tend to grow in dense clusters on the former and detach readily. Most cells are moderately adherent. Remove attached cells from the substrate with trypsin/EDTA. Cells will detach in 5-10 minutes. When resuscitated from a frozen ampoule cells may appear dead after a day but reattach and resume growth within 2-3 days. Cells grow best and are most adherent on a plastic substrate in medium at a pH of 6.9 - 7.2; they do not tolerate more alkaline pH well. Cultures hold well at high density with periodic medium changes.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service