Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V79

86041102, hamster lung, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

V79, 86041102, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

hamster lung

packaging

tube of 5 μg 86041102-DNA-5UG
pkg of vial of cells 86041102-1VL

growth mode

Adherent

karyotype

2n=22

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese lung male

Cell Line Description

Established after spontaneous transformation of cells isolated from the lung of a normal Chinese hamster (male). The G1 phase in these cells is either absent or very short. Consequently they have been used in studies of the G1 growth period. A clone called V79.4 is available (Sigma Catalogue number. 93010723). V79 is well established in toxicology studies. Stability of karyotype and morphology is making them suitable for gene toxicity assays with low background aberrations. The cells do not express endogenous cytochrome P450 and have been used for the insertion and expression of P450 from human, rat, mouse and fish.

Application

Growth factors and transformation studies, mutagenesis study
V79 has been used to detect structural and numerical chromosome damage by measuring the formation of micronuclei in interphase cells that have been through a mitotic division.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:12 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service