Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3X63Ag8.653 Cell Line from mouse

85011420, mouse blood, Not specified

Synonym(s):

Hybridoma cell line, Mouse cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P3X63Ag8.653 Cell Line from mouse, 85011420

biological source

mouse blood

description

Mouse BALB/c myeloma

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c myeloma

Cell Line Description

P3X63Ag8.653 is a non-Ig-secreting or synthesising subclone of P3X63.Ag8. Comparable to other myeloma lines used for fusions relating to growth properties, fusion frequency and stability of resulting hybrids.

Application

Fusion partner, growth factors and tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-5x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service