Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001598

Dutasteride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost-1-ene-17-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30F6N2O2
CAS Number:
Molecular Weight:
528.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dutasteride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=C[C@@]2(C)[C@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@H]4C(NC5=CC(C(F)(F)F)=CC=C5C(F)(F)F)=O)([H])N1

InChI

1S/C27H30F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,10,12-13,15-17,19,21H,4,6-9,11H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,21+,24-,25+/m0/s1

InChI key

JWJOTENAMICLJG-QWBYCMEYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dutasteride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Minoxidil British Pharmacopoeia (BP) Reference Standard

BP637

Minoxidil

Adriana C Vidal et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 23(12), 2936-2942 (2014-09-30)
Studies suggest that obesity is associated with lower risk of prostate cancer but more aggressive cancers. As obesity lowers PSA levels, these observations may be influenced by detection bias. We examined the association between obesity and risk of low- and
Matthias Oelke et al.
World journal of urology, 32(5), 1141-1147 (2014-06-07)
To assess the impact of dutasteride compared with placebo on nocturia in men with lower urinary tract symptoms suggestive of benign prostatic hyperplasia, using pooled data from dutasteride phase III studies. Nocturia was assessed using Question 7 of the International
Matthias Oelke et al.
World journal of urology, 32(5), 1133-1140 (2014-05-09)
The purpose of the study was to assess the impact of dutasteride plus tamsulosin combination therapy, compared with dutasteride or tamsulosin monotherapy, on nocturia in men with lower urinary tract symptoms suggestive of benign prostatic hyperplasia (LUTS/BPH) using data from
Gerald L Andriole et al.
Urology, 84(2), 393-399 (2014-06-12)
To explore explanations for the numerical imbalance of biopsy-detected Gleason 8-10 prostate cancers (PCa) diagnosed in years 3-4 in the dutasteride and placebo groups of the Reduction by Dutasteride of Prostate Cancer Events (REDUCE) study. REDUCE was a 4-year, randomized
Elahe A Mostaghel et al.
PloS one, 9(10), e111545-e111545 (2014-10-31)
Factors influencing differential responses of prostate tumors to androgen receptor (AR) axis-directed therapeutics are poorly understood, and predictors of treatment efficacy are needed. We hypothesized that the efficacy of inhibiting DHT ligand synthesis would associate with intra-tumoral androgen ratios indicative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service