Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V79-4

93010723, hamster lung, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

V79-4, 93010723

biological source

hamster lung

growth mode

Adherent

karyotype

2n = 22

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese lung

Cell Line Description

Derived from the lung tissue of a male Chinese hamster. The cells exhibit a high plating efficiency and a doubling time of 12-14 hours. Originally used in the discovery of X-ray induced damage and recovery in mammalian cells. This is a clone of cell line V79 (Sigma Catalogue number. 86041102).

Application

Mutagenesis and mammalian cell genetic studies

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 to 1:14 i.e. seeding at 3x10,000-1x100,000 cells/cm2; 5% CO2; 37°C using 0.25% trypsin/EDTA.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service