Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32222

Sigma-Aldrich

Dichloromethane

puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)

Synonym(s):

methylene chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
technique(s):
GC/GC: suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. ISO

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

1223 °F

contains

~25 mg/L amylene as stabilizer

expl. lim.

22 %

technique(s)

GC/GC: suitable

impurities

≤0.00002% free chlorine (Cl)
≤0.0005% formaldehyde
≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.01% chloroform (GC)
≤0.01% tetrachloromethane (GC)
≤0.01% water (Karl Fischer)
≤0.05% ethanol (GC)

refractive index

n20/D 1.4235-1.4245
n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.320-1.330 g/L at 20 °C
1.325 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 mg/kg

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

ClCCl

suitability

passes test for reaction against H2SO4

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. The apparent adsorption constant of the TiO2 loaded activated carbon for dichloromethane was found to be inversely proportional to the quantity of loaded TiO2.

Application

Dichloromethane may be used as a solvent for the preparation of stock solutions of carotenoids. It may be employed as a solvent in the synthesis of polylactide-based (PLA) thin films.

Other Notes

The article number 32222-4X2.5L will be discontinued. Please order the single bottle 32222-2.5L which is physically identical with the same exact specifications.
The article number 32222-4X2.5L-M will be discontinued. Please order the single bottle 32222-2.5L-M which is physically identical with the same exact specifications.
The article number 32222-6X1L will be discontinued. Please order the single bottle 32222-1L which is physically identical with the same exact specifications.
The article number 32222-6X1L-M will be discontinued. Please order the single bottle 32222-1L-M which is physically identical with the same exact specifications.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Re et al.
Free radical biology & medicine, 26(9-10), 1231-1237 (1999-06-25)
A method for the screening of antioxidant activity is reported as a decolorization assay applicable to both lipophilic and hydrophilic antioxidants, including flavonoids, hydroxycinnamates, carotenoids, and plasma antioxidants. The pre-formed radical monocation of 2,2'-azinobis-(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS*+) is generated by oxidation
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
Effect of activated carbon content in TiO2-loaded activated carbon on photodegradation behaviors of dichloromethane.
Torimoto T, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 103(1), 153-157 (1997)
Dipyridine-chromium (VI) oxide oxidation of alcohols in dichloromethane.
Collins JC, et al.
Tetrahedron Letters, 9(30), 3363-3366 (1968)
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service