Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

34856

Sigma-Aldrich

Dichloromethane

Dichloromethane

suitable for HPLC, ≥99.8%, contains amylene as stabilizer

Synonym(s):

methylene chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.04
Assay:
≥99.8%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

1223 °F

contains

amylene as stabilizer

expl. lim.

22 %

technique(s)

HPLC: suitable

impurities

free chlorine, in accordance
≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.02% water (Karl Fischer)

color

APHA: ≤10

transmittance

235 nm, ≥20%
240 nm, ≥50%
245 nm, ≥80%
260 nm, ≥98%

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

absorption

in accordance

application(s)

food and beverages

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml.[1] Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.[2]

Application

Dichloromethane may be used in ultrasonic baths for cleaning and protection of aluminum coating over atactic-PMMA (poly(methyl methacrylate)) for dielectric studies.[3]

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
  • The article number 34856-4X2.5L will be discontinued. Please order the single bottle 34856-2.5L which is physically identical with the same exact specifications.
  • The article number 34855-6X1L will be discontinued. Please order the single bottle 34856-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glassy dynamics and glass transition in thin polymer layers of PMMA deposited on different substrates.
Erber M, et al.
Macromolecules, 43(18), 7729-7733 (2010)
Effect of activated carbon content in TiO2-loaded activated carbon on photodegradation behaviors of dichloromethane.
Torimoto T, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 103(1), 153-157 (1997)
Dipyridine-chromium (VI) oxide oxidation of alcohols in dichloromethane.
Collins JC, et al.
Tetrahedron Letters, 9(30), 3363-3366 (1968)
K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service