Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNPLE-A012

Sigma-Aldrich

Reagent-cartridge Amide formation (for amine salts)

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.22

General description

Reagent-cartridge Amide formation (for amine salts) is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the amide coupling of carboxylic acids and amines. The cartridge contains all necessary reagents for amide coupling and purification.

Scale: up to 0.5 mmol

Compatible substrates: carboxylic acids, primary and secondary alkyl amine salts

Cartridge contents:
• Supported carbodiimide and Oxyma Pure (coupling reagents)
• SCX2 (for purification)
• Supported carbonate (acid scavenger)

Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
• Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
• Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
• Safety – all chemicals are contained in the system, minimizing the user’s exposure
• Set-up reaction in less than 5 minutes
• Small footprint
• Plug & Play system – simple installation
• Reaction temperatures from 25 °C – 80 °C
• Cartridges available for many different reactions
• Prefilled with all required reagents
• RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service