Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNPLE-BT002

Sigma-Aldrich

Reagent-cartridge Biotin tag-PEG2-NH2

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.22

Quality Level

parameter

up to 0.1 mmol scale

Looking for similar products? Visit Product Comparison Guide

General description

Reagent-cartridge Biotin tag-PEG2-NH2 is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the biotin tagging of aldehydes and ketones through a reductive amination. The cartridge contains all necessary reagents for linking the chosen aldehyde or ketone substrate to biotin.

Scale: up to 0.1 mmol
Linker: PEG2
Terminal functional group: amine
Compatible substrates: aldehyde, ketone

Cartridge contents:
  • Reducing agent (supported cyanoborohydride)
  • Base (supported triethylamine)
  • SCX (for purification)
  • Biotin linker

Related Cartridges:
  • SYNPLE-BT003Reagent-cartridge Biotin tag-PEG3-NH2
  • SYNPLE-BT004Reagent-cartridge Biotin tag-PEG4-NH2
  • SYNPLE-BT012Reagent-cartridge Biotin tag-PEG2-O-4-P-NH
  • SYNPLE-BT013Reagent-cartridge Biotin tag-PEG3-O-4-P-NH
  • SYNPLE-BT014Reagent-cartridge Biotin tag-PEG4-O-4-P-NH
  • SYNPLE-BT022Reagent-cartridge Biotin tag-PEG2-a-COOTfp
  • SYNPLE-BT023Reagent-cartridge Biotin tag-PEG3-a-COOTfp
  • SYNPLE-BT024Reagent-cartridge Biotin tag-PEG4-a-COOTfp
  • SYNPLE-BT032Reagent-cartridge Biotin tag-PEG2-COOTfp
  • SYNPLE-BT033Reagent-cartridge Biotin tag-PEG3-COOTfp
  • SYNPLE-BT034Reagent-cartridge Biotin tag-PEG4-COOTfp

Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
  • Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
  • Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety - all chemicals are contained in the system, minimizing the user′s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system - simple installation
  • Reaction temperatures from 25 °C - 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service