Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNPLE-A011

Sigma-Aldrich

Reagent-cartridge Amide formation

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352401
NACRES:
NA.22

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reagent-cartridge Amide formation is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the amide coupling of carboxylic acids and amines. The cartridge contains all necessary reagents for amide coupling and purification.

Scale: up to 0.5 mmol

Compatible substrates: carboxylic acids, primary and secondary alkyl amines

Cartridge contents:
  • Supported carbodiimide and Oxyma Pure (coupling reagents)
  • SCX2 (for purification)
  • Supported carbonate (acid scavenger)
Learn more about the Synple automated synthesis platform.


Features and Benefits of Synple Platform
  • Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
  • Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety – all chemicals are contained in the system, minimizing the user’s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system – simple installation
  • Reaction temperatures from 25 °C – 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagentsRFID chip containing optimized reaction protocol
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service