Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNPLE-P084

Sigma-Aldrich

Reagent-cartridge VHL-COOH-PEG4

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352401
NACRES:
NA.22

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI

1S/C28H38N4O7S/c1-17-24(40-16-30-17)19-7-5-18(6-8-19)14-29-26(37)21-13-20(33)15-32(21)27(38)25(28(2,3)4)31-22(34)9-11-39-12-10-23(35)36/h5-8,16,20-21,25,33H,9-15H2,1-4H3,(H,29,37)(H,31,34)(H,35,36)/t20-,21+,25-/m1/s1

Looking for similar products? Visit Product Comparison Guide

General description

Reagent-cartridge VHL-COOH-PEG4 is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the synthesis of molecules for targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. The cartridge contains all necessary reagents for linking the user selected amine to the carboxylic acid terminated linker through an amide coupling. This cartridge features a von Hippel-Lindau (VHL) recruiting ligand ((S,R,S)-AHPC) and PEG linker. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation.

Scale: up to 0.1 mmol

Linker: PEG4

E3 ligase target and ligand: von Hippel-Lindau (VHL), (S,R,S)-AHPC

Terminal functional group: carboxylic acid

Compatible substrates: amine

Cartridge contents:
  • Supported carbodiimide and Oxyma Pure (coupling reagents)
  • Supported carbonate (acid scavenger)
  • SCX2 (for purification)
  • Degrader building block linker reagent
Related Cartridges:
  • SYNPLE-P001 Reagent-cartridge CRBN-PEG1
  • SYNPLE-P002 Reagent-cartridge CRBN-PEG2
  • SYNPLE-P003 Reagent-cartridge CRBN-PEG3
  • SYNPLE-P004 Reagent-cartridge CRBN-PEG4
  • SYNPLE-P005 Reagent-cartridge CRBN-PEG5
  • SYNPLE-P022 Reagent-cartridge VHL-PEG2
  • SYNPLE-P023 Reagent-cartridge VHL-PEG3
  • SYNPLE-P024 Reagent-cartridge VHL-PEG4
  • SYNPLE-P025 Reagent-cartridge VHL-PEG5
  • SYNPLE-P072 Reagent-cartridge CRBN-COOH-PEG2
  • SYNPLE-P073 Reagent-cartridge CRBN-COOH-PEG3
  • SYNPLE-P074 Reagent-cartridge CRBN-COOH-PEG4
  • SYNPLE-P082 Reagent-cartridge VHL-COOH-PEG2
  • SYNPLE-P083 Reagent-cartridge VHL-COOH-PEG3
Learn more about the Synple automated synthesis platform.

Learn more about protein degradation.

Explore all our Protein Degrader Building Blocks.

Features and Benefits of Synple Platform
  • Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
  • Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety – all chemicals are contained in the system, minimizing the user’s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system – simple installation
  • Reaction temperatures from 25 °C – 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service