Quality Level
reaction suitability
reaction type: Reductive Aminations
parameter
up to 0.5 mmol scale
SMILES string
[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C
InChI key
HVPZHRHEHKZOOJ-TYBLODHISA-N
Related Categories
General description
Reagent-cartridge Reductive Amination is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the reductive amination of aldehydes and ketones. The cartridge contains all necessary reagents for condensation, reduction, and purification.
Scale: up to 0.5 mmol
Compatible substrates: aldehydes, ketones, primary amines, secondary amines
Cartridge contents:
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
Scale: up to 0.5 mmol
Compatible substrates: aldehydes, ketones, primary amines, secondary amines
Cartridge contents:
- Reducing agent (supported cyanoborohydride)
- Proton source (Phenylacetic acid)
- SCX (for purification)
- Supported benzaldehyde (amine scavenger)
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
- Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety - all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system - simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
Product of Synple Chem AG
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SYNPLE-R001-1EA | 04065266967920 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service