Skip to Content
MilliporeSigma
All Photos(1)

Documents

NISTRM8183

Omega-3 and Omega-6 Fatty acids in botanical oils

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.24

manufacturer/tradename

NIST®

format

matrix material

storage temp.

2-8°C

General description

Other Notes

Certified for the analytes listed below.
Octanoic Acid, Decanoic Acid, Dodecanoic Acid, Lauric Acid, Myristic Acid, Pentadecanoic Acid, Hexadecanoic Acid, Heptadecanoic Acid, Octadecanoic Acid , (Z)-9-Octadecenoic Acid, (E)-9-Octadecenoic Acid, (Z)-11-Octadecenoic Acid, (Z,Z)-9,12-Octadecadienoic Acid, (Z,Z,Z)-9,12,15-Octadecatrienoic Acid, (Z,Z,Z)-6,9,12-Octadecatrienoic Acid

Reference/Informational value is provided for the following analytes.
Eicosanoic Acid (C20:0), (Z)-11-Eicosenoic Acid, (Z,Z,Z,Z)-5,8,11,14-Eicosatetraenoic Acid, Heneicosanoic Acid, Docosanoic Acid, Tetracosanoic Acid, (Z)-15-Tetracosenoic Acid

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Kit Components Only

Product No.
Description

  • Seed oil - Borage

  • Seed oil - Evening Primrose

  • Seed oil - Flax

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service