Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2582

Powdered paint

NIST® SRM® 2582, nominal 200mg/kg lead

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 20 g

manufacturer/tradename

NIST®

application(s)

coatings

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Application

  • Enhancement of cultural heritage conservation techniques: Powdered paint was employed in non-destructive and non-invasive identification techniques for hydroxy lead-calcium phosphate solid solutions in cultural heritage materials, demonstrating its application in art restoration and conservation (Costantino et al., 2024).
  • Development of environmentally friendly building materials: Powdered paint was used in sustainable applications involving waste sludges from wastewater treatment plants, specifically in the construction industry to enhance the environmental sustainability of building materials (Stojmenovic et al., 2024).
  • Improvement of water-based paint formulations: A study assessed the impact of a sepiolite-based bio-pigment, infused with indigo extract, on the appearance and durability of water-based white primers, highlighting its significance in the development of more durable and aesthetically pleasing paint products (Calovi and Rossi, 2024).

Other Notes

Certified for the analytes listed below.
Lead (Pb)

Reference/Informational value is provided for the following analytes.
Alminium (Al), Calcium (Ca), Iron (Fe), Magnesium (Mg), Titanium (Ti), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service