Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2569

Lead paint films for children′s products

NIST® SRM® 2569

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 8 sheets

manufacturer/tradename

NIST®

application(s)

coatings

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in the evaluation of lead in paint on films or similar matrices using non-destructive, instrumental methods of analysis. SRM 2569 consists of three different paint films each containing a specified amount of lead (Pb) and each supported on polyester sheet. SRM 2569 is not intended for use in test method calibration for thickness and density. A unit of SRM 2569 consists of three coupons of paint on polyester and five coupons of uncoated polyester. The coupons are approximately 73 mm long by 52 mm wide. The paint films are cured lacquer having a specified thickness in the range of 20 μm to 45 μm. For more information, please refer to the COA and SDS.

SRM 2569_cert

SRM 2569_SDS

Other Notes

Certified for the analytes listed below.
Lead (Pb)

Reference/Informational value is provided for the following analytes.
Polyester substrate

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service