Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2574

Lead paint film for portable x-ray fluorescence analyzers

NIST® SRM® 2574, nominal 0.7 mg/cm2 (color code:  gold)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 1 +blank

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

coatings

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each SRM 2574 unit is a white polyester sheet about 7.6 cm wide, 10.2 cm long, and 0.2 mm thick. It is coated with a gold-colored paint layer of ~0.04 mm thick.



SRM 2574_cert

SRM 2574_SDS

Application

Lead paint film for building surfaces standard reference material (SRM) is intended to validate the results derived from portable,hand-held X-ray fluorescence analyzers for the testing of lead in paintcoatings applied to both interior and exterior building surfaces.

Features and Benefits

  • Available with a certificate documenting NIST-certified value of total lead areic mass in cured paint by isotope dilution inductively coupled plasma mass spectrometry (ID-ICP-MS).
  • Certified value is metrologically traceable to the International System of Units (SI) for mass and length which is expressed as milligrams per centimetersquared.
  • The NIST certificate is provided with expiration certificate, storage, handling, maintenance,and use instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service